Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Bromodiphenylmethane 97.0+%, TCI America™
SDP

Catalog No. B0586100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0586100G 100 g
B058625G 25 g
B0586500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0586100G Supplier TCI America Supplier No. B0586100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 776-74-9
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD00000134
InChI Key OQROAIRCEOBYJA-UHFFFAOYSA-N
Synonym bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
PubChem CID 236603
IUPAC Name [bromo(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Br

Specifications

Melting Point 39°C
Boiling Point 193°C
Quantity 100 g
UN Number 1770
Formula Weight 247.14
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material alpha-Bromodiphenylmethane
RTECSNumber PA5350000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.