Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha-Bungarotoxin Conjugates

Catalog No. B56130
Encompass_Preferred
Change view
Click to view available options
Label or Dye:
Alexa Fluor™ 488
Alexa Fluor™ 555
Alexa Fluor™ 594
Alexa Fluor™ 647
Alexa Fluor™ Plus 405
Biotin-XX
Tetramethylrhodamine
Unlabeled
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye
B56130 Alexa Fluor™ Plus 405
B13422 Alexa Fluor™ 488
B35451 Alexa Fluor™ 555
B13423 Alexa Fluor™ 594
B35450 Alexa Fluor™ 647
B1196 Biotin-XX
T1175 Tetramethylrhodamine
B1601 Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. B56130 Supplier Invitrogen™ Supplier No. B56130
  • $686.00
    $562.52 / Each of 1

    Save $123.48

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates.

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions. Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor.

Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

TRUSTED_SUSTAINABILITY

Specifications

Color Violet
Quantity 100 μg
Content And Storage Store at ≤-20°C, protect from light. Avoid freeze-thaw cycles.

Label or Dye Alexa Fluor™ Plus 405
Excitation/Emission 411/431 nm
Product Line Alexa Fluor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.