Learn More
Abcam Alpha-Calcitonin Gene Related Peptide, Human, CGRP Agonist, Solid, 500 ug
Description
- Molecular Weight: 3,789.3 Da
- Purity: >95%
- Form: Solid
- Constituents: 100% α-Calcitonin Gene Related Peptide (human)
- CAS Number: 90954-53-3
- Activity: CGRP agonist
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Calcitonin Gene Related Peptide (Human) |
| Shipping Condition | Ambient |
| Reactivity | Human |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 500 μg |
| For Use With (Application) | CGRP agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.