Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclodextrin, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600438 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-438 500 g Amber Glass Bottle
18-600-439 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-600-438 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1620500GMBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1620, 10016-20-3, C36H60O30

Spectrum™ Chemical alpha-Cyclodextrin , a polysaccharide, can be used as a dietary fiber. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.85
InChI Key HFHDHCJBZVLPGP-UHFFFAOYNA-N
IUPAC Name 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Water 10%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.