Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Alpha-Cyclodextrin, Non-Reducing Cyclic Saccharide, 5 g
Description
- Chemical structure: Non-reducing cyclic saccharide consisting of six α-1,4-linked D-glucopyranosyl units
- Molecular weight: 972.86 g/mol
- Production: Produced by the action of cyclodextrin glucosyltransferase (CGTase, EC 2.4.1.19) on hydrolyzed starch
- Applications: Useful for the solubilization of nonpolar macromolecules such as fatty acids, lipids, and cholesterol for cell culture
- Composition: 100% α-Cyclodextrin hydrate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Form | Powder |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Fatty Acids, Lipids, Cholesterol |
| Formulation | 100% α-Cyclodextrin hydrate |
| Quantity | 5 g |
| For Use With (Application) | Cell culture applications, solubilization of nonpolar macromolecules |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?