Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Cyclopentyl-DL-mandelic Acid 98.0+%, TCI America™
SDP

Catalog No. C239425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C239425G 25 g
C23945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C239425G Supplier TCI America Supplier No. C239425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 427-49-6
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
MDL Number MFCD00019296
InChI Key WFLUEQCOAQCQLP-UHFFFAOYSA-N
Synonym alpha-cyclopentylmandelic acid, cyclopentylphenylglycolic acid, cyclopentyl hydroxy phenylacetic acid, cyclopentyl-hydroxy-phenyl-acetic acid, cyclopentyl mandelic acid, alpha-cyclopentyl-dl-mandelic acid, a-cyclo pentyl mandelic acid, alpha-cyclopentylmandelicacid, 2-rs-cyclopentyl-2-hydroxy-2-phenylacetic acid alpha-cyclopentylmandelic acid, cyclopentylmandelic acid
PubChem CID 98283
IUPAC Name 2-cyclopentyl-2-hydroxy-2-phenylacetic acid
SMILES C1CCC(C1)C(C2=CC=CC=C2)(C(=O)O)O

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 25 g
Formula Weight 220.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyclopentyl-DL-mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.