Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-D-Glucose-1-phosphate dipotassium salt hydrate, 98%, Thermo Scientific™

Catalog No. p-7043197
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6615909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6615909 Supplier Thermo Scientific Chemicals Supplier No. J6615909

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A α-anomeric form of glucose which contains a phosphate group on the primary carbon. Used as a substance for the estimation of phosphorylases.

Solubility
Soluble in water

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1233690-06-6
Molecular Formula C6H17K2O11P
Molecular Weight (g/mol) 374.361
MDL Number MFCD00149435
InChI Key RZULLROGARPECB-QMKHLHGBSA-N
Synonym alpha-d-glucopyranose, 1-dihydrogen phosphate , dipotassium salt, dihydrate, alpha-d-glucopyranose 1-phosphate dipotassium salt hydrate, alpha-d-glucose 1-phosphate dipotassium salt dihydrate, alpha-glucose-1-phosphate dipotassium dihydrate, alpha-glucose-1-phosphate dipotassium salt dihydrate mi, a-d-glucose-1-phosphate dipotassium salt hydrate, alpha-d-glucose-1-phosphate dipotassium salt hydrate, dipotassium dihydrate ?-d-glucopyranosyl phosphate, dipotassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphate dihydrate, potassium 2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl phosphate dihydrate
PubChem CID 131855785
IUPAC Name potassium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate;dihydrate
SMILES C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.O.O.[K].[K]

Specifications

Color White
Odor Odorless
Quantity 10 g
Merck Index 14,4461
Solubility Information Soluble in water
Formula Weight 336.32 (Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material alpha-D-Glucose-1-phosphate dipotassium salt hydrate
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.