Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals alpha-D(+)-Glucose, 99+%, anhydrous

Catalog No. AC170080010
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC170080010 1 kg
AC170080025 2.5 kg
AC170080250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC170080010 Supplier Thermo Scientific Chemicals Supplier No. 170080010
  • $61.50
    $49.65 / Each of 1

    Save $11.85

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 492-62-6 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-62-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063774
InChI Key WQZGKKKJIJFFOK-DVKNGEFBSA-N
Synonym alpha-d-glucose, alpha-d-glucopyranose, alpha-dextrose, alpha-glucose, glucopyranose, alpha-d, alpha-d-glc, unii-5j5i9eb41e, glucoses, grape sugar, d-gluco-hexose
PubChem CID 79025
ChEBI CHEBI:17925
IUPAC Name (2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)O)O)O)O)O

Specifications

Acidity 0.15mL 0.1N NaOH/6g max.
CAS Min % 99.0
CAS Max % 100.0
Melting Point 156.0°C to 158.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 31, 83
Merck Index 14, 4459
Specific Rotation Condition + 52.90 (20.00°C c=10,water,ammonia)
Specific Rotation + 52.90
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 180.16
Percent Purity 99+%
Related Substances 0.15% max. (Fructose)
Physical Form Fine Crystalline Powder
Chemical Name or Material α-D(+)-Glucose, Anhydrous
Show More Show Less
EINECSNumber 207-757-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.