Learn More
CPC Scientific H-Ala-Phe-Thr-Cys-His-Cys-Arg-Arg-Ser-Cys-Tyr-Ser-Thr-Glu-Tyr-Ser-Tyr-Gly-Thr-Cys-Thr-Val-Met-Gly-Ile-Asn-His-Arg-Phe-Cys-Cys-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific DEFS008C
Encompass
This peptide contain 6 conserved disulfide-linked cysteines. In vitro, it has a prominent antimicrobial activities against bacteria, fungi, and certain enveloped viruses. In addition, human and rabbit defensins exert potent cytotoxicity in vitro against various mammalian tumor cells.SEQUENCE: H-Ala-Phe-Thr-Cys-His-Cys-Arg-Arg-Ser-Cys-Tyr-Ser-Thr-Glu-Tyr-Ser-Tyr-Gly-Thr-Cys-Thr-Val-Met-Gly-Ile-Asn-His-Arg-Phe-Cys-Cys-Leu-OH (trifluoroacetate salt)(Cys4 and Cys31/Cys6 and Cys20/Cys10 and Cys30 Disulfide Bridges)ONE-LETTER SEQUENCE: AFTCHCRRSCYSTEYSYGTCTVMGINHRFCCL (Cys4 and Cys31/Cys6 and Cys20/Cys10 and Cys30 Disulfide Bridges)MOLECULAR FORMULA: C156H228N46O46S7MOLECULAR WEIGHT: 3708.27STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [136661-76-2]SYNONYMS: -Defensin 6RESEARCH AREA: AntimicrobialCATEGORY: Defensins REFERENCES:P.A.Raj et al., Biochem. J., 347, 633 (
Catalog No. 50-210-9198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.