Learn More
Novus Biologicals™ alpha-Fetoprotein/AFP Recombinant Protein Antigen
Shop All Novus Biologicals Products

Specifications
Specifications
| Gene ID (Entrez) | 174 |
| Species | Human |
| Purity | > 80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Quantity | 0.1 mL |
| Research Category | Cancer, Cancer Stem Cells, Developmental Biology, Stem Cell Markers, Tumor Biomarkers |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-55068. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
| Storage Buffer | PBS and 1M Urea, pH 7.4. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.