Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Alpha Galactosidase Activity Assay Kit, Fluorometric, 2000 Tests

Catalog No. 502586147
Encompass_Preferred
Change view
Click to view available options
Quantity:
2,000 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-6147 2,000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-6147 Supplier Abcam Supplier No. AB2397162000TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

Quantitative fluorometric assay for measuring alpha galactosidase activity in various biological samples.

  • Detection method: Fluorescent
  • Applications: Enzyme activity assay
  • Sample types: Saliva, Urine, Tissue Homogenate, Serum, Cell Lysate
  • Assay time: 2 hr
  • Quantitative assay
  • Includes positive control
  • Compatible with microplate readers

Specifications

Target Molecule Alpha Galactosidase
No. of Reactions 2000 Tests
For Use With (Equipment) Microplate Reader
Detection Location In-Solution Detection
Format Kit
Detection Method Fluorescence
Sample Type Saliva, Urine, Tissue Homogenate, Serum, Cell Lysate
Quantity 2,000 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.