Learn More
Invitrogen™ alpha Galactosidase Polyclonal Antibody

Description
PA1-9528 detects Alpha Galactosidase in human samples. PA1-9528 has been successfully used in Western blot and ELISA procedures. The PA1-9528 immunogen is a mix of synthetic peptides corresponding to residues 55-64 and 396-407 of human alpha Galactosidase. Store at 4ºC for one week, and -20ºC for long term storage.
Specifications
Specifications
| Antigen | alpha Galactosidase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GLA |
| Gene Accession No. | P06280 |
| Gene Alias | Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; Melibiase |
| Gene Symbols | GLA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.