Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Galactosylceramide (alpha-Gal-Cer, KRN7000), MP Biomedicals™

Catalog No. MP218331601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218331601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218331601 Supplier MP Biomedicals, Inc Supplier No. 0218331601
Only null left
Add to Cart
Add to Cart

Used in cancer research as an antitumor compound to induce a NKT immuno response.

  • An immuno stimulator and that stimulates NKT, Natural Killer T cells
  • Uses a specific ligand of lipid binding MHC class I- Like protein CD1d in mouse and human natural killer T cells
  • It is a potent antitumor compound that protects against LPS - Induced Shocked
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Melting Point 189°C to 190°C
Boiling Point 189°C
Color White
Purity >96
Molecular Weight (g/mol) 858.3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.