Learn More
CPC Scientific H-Phe-His-Leu-Leu-Arg-Glu-Met-Leu-Glu-Met-Ala-Lys-Ala-Glu-Gln-Glu-Ala-Glu-Gln-Ala-Ala-Leu-Asn-Arg-Leu-Leu-Leu-Glu-Glu-Ala-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CRFS005A
Encompass
ONE-LETTER SEQUENCE: FHLLREMLEMAKAEQEAEQAALNRLLLEEA-NH2MOLECULAR FORMULA: C152H251N43O47S2MOLECULAR WEIGHT:3497.06STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [158535-55-8], SYNONYMS: α-Helical CRF (12-41)RESEARCH AREA: Hormonal
Catalog No. 50-210-9167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.