Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-II Spectrin Recombinant Rabbit Monoclonal Antibody (19H12L58)

Catalog No. 703784
Encompass_Preferred
missing translation for 'changeView'
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-378-4 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 70-378-4 Supplier Invitrogen™ missing translation for 'supplierNo' 703784
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Bovine, Horse. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

SPTAN1 (spectrin alpha chain, non-erythrocytic 1) is a part of the spectrin family of filamentous cytoskeletal proteins. They function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. SPTAN1 has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-II Spectrin
Applications Western Blot
Classification Recombinant Monoclonal
Clone 19H12L58
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene SPTAN1
Gene Accession No. Q13813
Gene Alias (ALPHA)II-SPECTRIN; 2610027H02Rik; A2a; a--fodrin; alpha; alpha II spectrin; alpha-fodrin; alpha-II spectrin; alpha-spectrin; alpha-spectrin 2; alpha-spectrin 2, brain; brain; brain alpha-spectrin; EIEE5; FLJ44613; Fodrin alpha chain; inhibitory protein factor; IPF; NEAS; noerythroid alpha-spectrin 2; Non erythrocytic spectrin alpha; non-erythrocytic 1 (alpha-fodrin); non-erythroid alpha chain; nonerythroid alpha-spectrin; nonerythroid alpha-spectrin 2; putative alpha-spectrin; SPECA; Spectrin; Spectrin alpha; spectrin alpha chain; Spectrin alpha chain brain; spectrin alpha chain, brain; spectrin alpha chain, non-erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; Spna2; Spna-2; Spta2; Sptan1
Gene Symbols SPTAN1
Host Species Rabbit
Immunogen Protein corresponding to human SPTAN1 [aa2330-aa2472].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6709
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.