Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Internexin Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670274 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-274 Supplier Invitrogen™ Supplier No. PA110017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western blot, a prominent band at ~66 kDa is observed, as well as smaller lower bands which are apparently degradation products. A minor band at ~150 kDa is also seen, resulting from dimerization of alpha-internexin.

alpha-Internexin is a Class IV intermediate filament subunit expressed very early in neuronal development. It is related to but distinct from the better known neurofilament triplet proteins, NF-L, NF-M and NF-H, having similar protein sequence motifs and a similar intron organization. Many classes of mature neurons contain a-internexin in addition to NF-L, NF-M and NF-H. In some mature neurons a-internexin is the only neurofilament subunit expressed. Antibodies to internexin are therefore unique probes to study and classify neuronal types and follow their processes in sections and in tissue culture. In addition the very early developmental expression of a-internexin means its presence is an early and convenient diagnostic feature of neuronal progenitors cells and other cell committed to the neuronal lineage. The use of antibodies to this protein in the study of brain tumors has not been examined to date, but is likely to be of interest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Internexin
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 5mM sodium azide
Gene INA
Gene Accession No. P23565, P46660, Q08DH7, Q16352
Gene Alias 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1
Gene Symbols INA
Host Species Rabbit
Immunogen Purified, recombinant rat alpha-internexin, expressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100059769, 100153400, 226180, 24503, 532236, 9118
Target Species Human, Mouse, Rat, Bovine, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.