Learn More
Invitrogen™ alpha Internexin Recombinant Rabbit Monoclonal Antibody (5E5Z3)

Description
Positive Samples: U-87MG, Mouse brain, Rat brain Immunogen sequence: EVAGYQDSIG QLENDLRNTK SEMARHLREY QDLLNVKMAL DIEIAAYRKL LEGEETRFST SGLSISGLNP LPNPSYLLPP RILSATTSKV SSTGLSLKKE E.
Specifications
Specifications
| Antigen | alpha Internexin |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 5E5Z3 |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | INA |
| Gene Accession No. | P23565, P46660, Q16352 |
| Gene Alias | 66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.