Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Ketoglutaric acid disodium salt dihydrate, 99%

Catalog No. AAJ6315422
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6315422 100 g
AAJ6315414 25 g
AAJ6315430 250 g
AAJ6315403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6315422 Supplier Thermo Scientific Chemicals Supplier No. J6315422
Add to Cart

CAS: 305-72-6 | C5H4Na2O5 | 190.062 g/mol

alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Ketoglutaric acid disodium salt dihydrate is used as pharmaceutical intermediate. α-Ketoglutaric acid disodium salt dihydrate is suitable reagent used in a study to investigate the role of nitric oxide (NO) in mast cell physiology and biochemistry.

Solubility
Soluble in water (1 M).

Notes
Hygroscopic. Store under inert gas. Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-72-6
Molecular Formula C5H4Na2O5
Molecular Weight (g/mol) 190.062
MDL Number MFCD00150702
InChI Key YBGBJYVHJTVUSL-UHFFFAOYSA-L
Synonym disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
PubChem CID 31040
IUPAC Name disodium;2-oxopentanedioate
SMILES C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (1 M).
Formula Weight 226.09 (190.06 Anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material alpha-Ketoglutaric acid disodium salt dihydrate
EINECSNumber 206-167-8
RTECSNumber SA0454700
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.