Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alpha-Macroglobulin (Mouse) ELISA Kit

Catalog No. 89032274 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-274 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-274 Supplier Abnova™ Supplier No. KA4820
Add to Cart
Add to Cart

Enzyme Immunoassay for the Quantitative Determination of Mouse α-Macroglobulin in Serum or Plasma

  • For quantitative measurement of alpha-macroglobulin in plasma and serum

Specifications

Calibration Range 3.9 to 125 ng/mL
Description Alpha-Macroglobulin (Mouse) ELISA Kit is for the quantitative measurement of alpha-macroglobulin in plasma and serum.
Conjugate HRP
Detection Method Colorimetric
Product Type Alpha-Macroglobulin (Mouse) ELISA Kit
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Application) Quantification
Kit Contents Anti-mouse α-macroglobulin antibody coated plate: 96 wells (12 detachable strips of 8), Lyophilized Reference standard: Diluent: 30mL, Enzyme Conjugate Reagent: 11mL, 20x Wash Solution: 50mL, TMB Reagent (One-step): 11mL, Stop Solution (1N HCl): 11mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at 4°C.
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.