Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Mangostin 97.0+%, TCI America™
SDP

Catalog No. M2793100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2793100MG 100 mg
M279310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2793100MG Supplier TCI America Supplier No. M2793100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6147-11-1
Molecular Formula C24H26O6
Molecular Weight (g/mol) 410.47
MDL Number MFCD00135200
InChI Key GNRIZKKCNOBBMO-UHFFFAOYSA-N
PubChem CID 5281650
ChEBI CHEBI:67547
IUPAC Name 1,3,6-trihydroxy-7-methoxy-2,8-bis(3-methylbut-2-enyl)xanthen-9-one
SMILES CC(=CCC1=C(C=C2C(=C1O)C(=O)C3=C(C(=C(C=C3O2)O)OC)CC=C(C)C)O)C

Specifications

Melting Point 180°C
Color Yellow
Quantity 100 mg
Formula Weight 410.47
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Mangostin
RTECSNumber ZD6122420
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.