Learn More
Bachem Mating Factor a Acetate
SDP
Supplier:  Bachem 4047325.0001
Encompass_Preferred
Mating Factor a Acetate, H-Trp-His-Trp-Leu-Gln-Leu-Lys-Pro-Gly-Gln-Pro-Met-Tyr-OH, 59401-28-4, C82H114N20O17S, Mr 1684.01, 1mg. alpha-Factor, a-Factor, Pheromone a-Factor, a-Mating Factor, yeast. Alpha-mating factor, a 13-amino acid pheromone secreted by haploid a-cells of Saccharomyces cerevisiae, binds to Ste2p, a transmembrane G-protein-coupled receptor present on haploid a-cells, and activates a signal transduction pathway required for conjugation and mating.
Catalog No. 50-194-7485
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.