Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha N-Catenin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103713 Supplier Thermo Scientific Supplier No. PA5103713
Only null left
Add to Cart
Add to Cart

alpha N-Catenin Polyclonal Antibody

alpha N-Catenin Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha N-Catenin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTNNA2
Gene Accession No. P26232, Q61301
Gene Alias AI481747; alpha N-catenin; alpha(N)-catenin; alpha-catenin-related protein; alpha-N-catenin; cadherin associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAPR; CAP-R; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 2; catenin alpha 2; catenin alpha-2; Catna; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA2; CTNR; DKFZp686H02198; OTTHUMP00000216460; OTTHUMP00000216462
Gene Symbols CTNNA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTNNA2(Accession P26232), corresponding to amino acid residues T21-F71.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12386, 1496
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.