Learn More
Eisco™ Alpha Scattering Apparatus
SCI_ED
Catalog No. S27427 Shop All Eisco Products
Change view
Click to view available options
For Use With (Application):
Demonstrating Rutherford Scattering
Catalog No. For Use With (Application)
S27427 Demonstrating Rutherford Scattering
1 options
Catalog No. S27427 Supplier Eisco™ Supplier No. ALPHA14
Add to Cart
Add to Cart
For demonstrating Rutherford scattering by means of a gravitational analogue of inverse square law repulsion
  • Apparatus comprises a Spun aluminum Hill, (60 x 280 mm), 2 Steel Balls (12.5, 19 mm), and a Wooden Launching Ramp (300 x 48 x 105 mm)
  • Shape has been determined by varying its height above the base line by an amount based upon the reciprocal of the radius
  • Launching ramp has a guide groove down its curved front face
  • Ramp includes index marks at 40, 60, 80 and 100 mm above the base to give a variety of known launching heights
Specifications
Product Type Alpha Scattering Apparatus
For Use With (Application) Demonstrating Rutherford Scattering
Includes Spun Aluminum Hill (1), Wooden Launching Ramp (1), Steel Balls (2)
Diameter (Metric) 280 mm (Hill)