Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha SMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501736008 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6008 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6008 Supplier Proteintech Group Inc Supplier No. 551351AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody stains the aorta only.

Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predomitly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.

Specifications

Antigen Alpha SMA
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACTA2
Gene Accession No. P62736, P62737, P62738
Gene Alias a SMA, AAT6, ACTA2, ACTA2 Smooth muscle Actin, ACTA2 Smooth muscle Actin Specific, Actin, aortic smooth muscle, Alpha actin 2, Alpha SMA, Alpha SMA Specific, a-SMA, smooth muscle actin, alpha SMA
Gene Symbols ACTA2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11475, 59, 81633
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.