Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Synuclein Monoclonal Antibody (4F1), PerCP
GREENER_CHOICE

Catalog No. PIMA545848
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545848 Supplier Invitrogen™ Supplier No. MA545848
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45848 was sufficient for detection of Alpha Synuclein in 15 μg of human brain cell lysate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Binds multiple linear epitopes within human alpha synuclein: 21-70 aa, 61-90 aa, and 101-140 aa, determined using Dot blot and ELISA with corresponding peptides. Detects approximately 14 kDa band corresponding to Alpha synuclein. Band at approximately 40 kDa is a trimer.

Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. These proteins are abundant in the brain and play a role in regulating synaptic transmission and membrane trafficking. Alpha- and beta-synuclein have been shown to selectively inhibit phospholipase D2, suggesting that they may also be involved in signaling pathways. Defects in the SNCA gene, which encodes alpha-synuclein, have been implicated in the pathogenesis of Parkinson's disease. Additionally, alpha-synuclein peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Multiple alternatively spliced transcripts encoding different isoforms have been identified for this gene. Diseases associated with SNCA include Parkinson Disease 1, Autosomal Dominant and Dementia, Lewy Body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Synuclein
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4F1
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SNCA
Gene Accession No. O55042, P37377, P37840
Gene Alias Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor)
Gene Symbols SNCA
Host Species Mouse
Immunogen Mouse alpha synuclein aggregate.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20617, 29219, 6622
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.