Learn More
Invitrogen™ alpha Tubulin Monoclonal Antibody (DM1A), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Description
Description: The monoclonal antibody DM1a recognizes the 50 kDa cytoskeletal protein alpha tubulin in a variety of species (human, mouse, rat, monkey, dog, pig, bovine, goat, hamster, guinea pig, kangaroo, amphibians, sea urchin, yeast and tobacco plants). Tubulin, the major component of microtubules, is a dimeric protein consisting of an alpha and beta subunit. Tubulin is a GTP-binding protein that can be modified by phosphorylation and acetylation resulting in assembly (polymerization) or disassembly (depolymerization). The dynamic nature of microtubules is most evident in the mitotic apparatus. The DM1a antibody recognizes the C-terminal end of the alpha tubulin isoform (amino acids 426-430). Applications Reported: This DM1A antibody has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: This DM1A antibody has been tested by immunocytochemistry of adherent cells. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | alpha Tubulin |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | DM1A |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | TUBAL3 |
| Gene Accession No. | A6NHL2, P02550, P05213, P0DPH7, P68363, P68366, P68368, P68369, P68370, P68373, Q2XVP4, Q3KRE8, Q3UX10, Q5XIF6, Q6AYZ1, Q6P9V9, Q6PEY2, Q71U36, Q9BQE3, Q9BVA1, Q9CWF2 |
| Gene Alias | 1t; a1t; ALPHA 84C; alpha tubulin; alpha tubulin 84B; alpha[[1]] tubulin; alpha[[1]]-tubulin; alpha1; alpha1 tubulin; alpha1t; alpha1tub; alpha1tub84B; alpha1-tubulin; alpha84B; alphaT; alphat-1; alphatub; alpha-tub; alphaTub1; alphatub84; alphaTUB84B; alpha-tub84B; alphaTub84B-PA; alphatubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 2; Alpha-tubulin 3; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; Alpha-tubulin 3E; alpha-tubulin 4; Alpha-tubulin 6; alphaTubulin 84B; alpha-tubulin 84B; alpha-Tubulin at 84B; alpha-tubulin I; Alpha-tubulin II; alpha-tubulin III; Alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-2; Alpha-tubulin isotype M-alpha-4; alpha-tubulin isotype M-alpha-6; alpha-tubulin TUB1; alpha-tubulin ubiquitous; alphaTubulin84B; Alpha-Tubulin84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; AT25469p; aTub84B; a-tub84B; bA408E5.3; B-ALPHA-1; bcm948; BEST:LD32507; cb944; CG1913; CG1913-PA; chr3R:2914276..2914416; clone 1.59; clone 2.30; Detyrosinated tubulin alpha-1A chain; Detyrosinated tubulin alpha-1B chain; Detyrosinated tubulin alpha-1C chain; Detyrosinated tubulin alpha-3 chain; Detyrosinated tubulin alpha-3C chain; Detyrosinated tubulin alpha-3D chain; Detyrosinated tubulin alpha-3E chain; DM1alpha; Dmel\CG1913; Dmel_CG1913; DTA1; fb22g06; FLJ30169; H2 ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub2; I79_019110; K-ALPHA-1; l(3)84Bd; l(3)g3; lis3; LOC100726777; LOC100731885; LOC100857858; LOC101118307; LOC106557476; M[a]4; M[a]6; ms(3)nc33; nc33; RGD1565476; similar to alpha-tubulin isoform 1; similar to tubulin alpha 1; T; Talpha1; Talpha1 alpha-tubulin; TBA1_DROME; Testis-specific alpha-tubulin; Tub; Tub 84B; TUB1; Tub1C; Tub84B; TUBA1; Tuba-1; tuba1 protein; TUBA1/3/4; Tuba1a; tuba1a.L; tuba1a-a; tuba1a-b; TUBA1B; TUBA1C; tuba1l; tuba1l2; TUBA2; TUBA3; TUBA3C; TUBA3D; TUBA3E; Tuba4; Tuba4a; TUBA5; Tuba6; tuba8.S; tubA84B; tubal2; Tubalpha1; tubulin; tubulin 1 alpha-chain; tubulin 1alpha; tubulin alpha; tubulin alpha 1a; tubulin alpha 1a L homeolog; tubulin alpha 1b; tubulin alpha 1c; tubulin alpha 3; tubulin alpha 3c; tubulin alpha 3e; tubulin alpha 4a; tubulin alpha 6; tubulin alpha 8 S homeolog; tubulin alpha1; tubulin alpha-1 chain; tubulin alpha1 promoter; Tubulin alpha-1A chain; tubulin alpha-1B chain; Tubulin alpha-1C chain; tubulin alpha-1C chain-like; tubulin alpha-2 chain; Tubulin alpha-3 chain; Tubulin alpha-3C chain; tubulin alpha-3C/D chain; Tubulin alpha-3D chain; Tubulin alpha-3E chain; tubulin alpha-4 chain; Tubulin alpha-4A chain; Tubulin alpha-5 chain; tubulin alpha-5 chain-like; tubulin alpha-6 chain; tubulin alpha-8 chain; tubulin alpha-ubiquitous chain; tubulin B-alpha-1; Tubulin H2-alpha; tubulin K-alpha-1; tubulin(alpha1); tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1, like; tubulin, alpha 1, like 2; tubulin, alpha 1a; tubulin, alpha 1b; tubulin, alpha 1c; tubulin, alpha 2; tubulin, alpha 3c; tubulin, alpha 3e; tubulin, alpha 4; tubulin, alpha 4a; tubulin, alpha 5; tubulin, alpha 6; tubulin, alpha, brain-specific; tubulin, alpha, ubiquitous; tubulin1alpha; tubulin84B; Tubulin-alpha1; Unknown (protein for MGC:133894); unnamed protein product; wu:fb22g06; wu:fb37a10; XELAEV_18019849mg; XELAEV_18020901mg; YML085C |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.