Learn More
Description
Recommended positive controls: Jurkat, Raji, 293T, A431, HeLa, HepG2, H1299, HCT116, MCF-7, NT2D1, PC-3, U87-MG, NIH-3T3, mouse brain, rat brain, Drosophila. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (98%), Pig (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TUBA1A |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.14 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | TUBA1A |
| Gene Accession No. | P68369, P68370, Q71U36 |
| Gene Alias | alpha 1a antibody; alpha 3 antibody; alpha-tubulin; alpha-tubulin 1; Alpha-tubulin 3; alpha-tubulin 3 antibody; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1; B-ALPHA-1 antibody; brain-specific antibody; Detyrosinated tubulin alpha-1A chain; FLJ25113 antibody; FLJ30169; H2 ALPHA; H2ALPHA; H2-ALPHA; hum-a-tub1; hum-a-tub1 antibody; hum-a-tub2; hum-a-tub2 antibody; LIS3; LIS3 antibody; LOC106557476; M40; MGC117247; MGC16435; OK/SW cl.56; OK/SWcl.56; OK/SW-cl.56; Talpha1 alpha-tubulin; TUBA1; Tuba-1; Tuba1a; TUBA1A antibody; TUBA3; TUBA3 antibody; TUBA6; TUBB1; TUBB5; tubulin alpha 1a; tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-1A chain antibody; tubulin alpha-3 chain; tubulin alpha-3 chain antibody; Tubulin B-alpha-1; tubulin B-alpha-1 antibody; tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 1A; tubulin, alpha 6; tubulin, alpha, brain-specific; unnamed protein product |
| Gene Symbols | TUBA1A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
