Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alphacel Hydrolyzed, MP Biomedicals™

Catalog No. MP290045405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
100 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290045405 5 lb.
MP290045425 25 lb.
MP290045480 100 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP290045405 Supplier MP Biomedicals, Inc Supplier No. 0290045405
Add to Cart
Add to Cart

Alphacel Hydrolyzed, White Powder

  • Alphacel is a polysaccharide composed of long chains of β(1,4) linked D-glucose units.
  • Cellulose is used in a wide range of applications that require a structural back-bone.
  • It is used in the preparation of chromatographic structural media, in gels for processes such as electrophoresis and ionic separations and as a substrate to identify and study cellulosic systems
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
MDL Number MFCD00081512
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N

Specifications

Color White
pH 5.5 to 7.0
Physical Form Powder
Quantity 5 lb.
RTECSNumber FJ5691460
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.