CAS RN 9004-34-6
Cellulose microcrystalline, average particle size 50 μm
CAS: 9004-34-6 MDL Number: MFCD00081512
Thermo Scientific Chemicals Cellulose, microcrystalline
CAS: 9004-34-6 MDL Number: MFCD00081512
Microcrystalline Cellulose, NF, BP, JP, Spectrum™ Chemical
CAS: 9004-34-6 Molecular Formula: C14H26O11 Molecular Weight (g/mol): 370.35 InChI Key: PTHCMJGKKRQCBF-UHFFFAOYNA-N IUPAC Name: 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol SMILES: COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
Microcrystalline Cellulose, NF, Spectrum™ Chemical
CAS: 9004-34-6 Molecular Formula: C14H26O11 Molecular Weight (g/mol): 370.35 InChI Key: PTHCMJGKKRQCBF-UHFFFAOYNA-N IUPAC Name: 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol SMILES: COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O
Microcrystallline cellulose powder, 98.6%, MP Biomedicals™
CAS: 9004-34-6 Molecular Weight (g/mol): 162.14
Cellulose powder, microcrystallline, >97%, MP Biomedicals™
CAS: 9004-34-6 Molecular Formula: (C6H10O5)n
Alphacel Non-Nutritive Bulk, MP Biomedicals
CAS: 9004-34-6
Alphacel Hydrolyzed, MP Biomedicals™
CAS: 9004-34-6 Molecular Formula: C14H26O11 Molecular Weight (g/mol): 370.35 MDL Number: MFCD00081512 InChI Key: PTHCMJGKKRQCBF-UHFFFAOYNA-N
Baker-flex™, Cellulose F, J.T. Baker™
CAS: 9004-34-6
Baker-flex™, Cellulose Microcrystalline, J.T. Baker™
CAS: 9004-34-6
Baker-flex™, Cellulose PEI-F, J.T. Baker™
CAS: 9004-34-6
Baker-flex™, Cellulose PEI, J.T. Baker™
CAS: 9004-34-6
- 1
- 2