Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alphacel Non-Nutritive Bulk, MP Biomedicals

Catalog No. ICN90045380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
100 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90045380 100 lb.
ICN90045305 5 lb.
ICN90045325 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN90045380 Supplier MP Biomedicals, Inc Supplier No. 0290045380
Add to Cart
Add to Cart

Composed of finely ground cellulose for addition to animal diets to supply bulk with minimum nutritional value

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6

Specifications

Quantity 100 lb.
Chemical Name or Material Alphacel Non-Nutritive Bulk
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.