Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ AlphaGuard™ Coveralls
BIOPHARMA

Catalog No. 191301502K Shop All AlphaProTech Products
Change view
Click to view available options
Color:
Blue
White
Description:
Elastic wrist, ankle and back
Elastic wrist, ankle and back, AquaTrak™ boots
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8X-Large
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description Size
19-130-1502K White Elastic wrist, ankle and back 8X-Large
19-130-1503A Blue Elastic wrist, ankle and back Small
19-130-1503B Blue Elastic wrist, ankle and back Medium
19-130-1503C Blue Elastic wrist, ankle and back Large
19-130-1503D Blue Elastic wrist, ankle and back X-Large
19-130-1503E Blue Elastic wrist, ankle and back 2X-Large
19-130-1503F Blue Elastic wrist, ankle and back 3X-Large
19-130-1503G Blue Elastic wrist, ankle and back 4X-Large
19-169-4011 Blue Elastic wrist, ankle and back 5X-Large
19-169-4026 Blue Elastic wrist, ankle and back 6X-Large
19-130-1502B White Elastic wrist, ankle and back Medium
19-130-1502C White Elastic wrist, ankle and back Large
19-130-1502D White Elastic wrist, ankle and back X-Large
19-130-1502E White Elastic wrist, ankle and back 2X-Large
19-130-1502F White Elastic wrist, ankle and back 3X-Large
19-130-1502G White Elastic wrist, ankle and back 4X-Large
19-130-1502H White Elastic wrist, ankle and back 5X-Large
19-130-1502I White Elastic wrist, ankle and back 6X-Large
19-125-604 White Elastic wrist, ankle and back, AquaTrak™ boots Small
19-125-600 White Elastic wrist, ankle and back, AquaTrak™ boots Medium
19-125-605 White Elastic wrist, ankle and back, AquaTrak™ boots Large
19-125-601 White Elastic wrist, ankle and back, AquaTrak™ boots X-Large
19-125-602 White Elastic wrist, ankle and back, AquaTrak™ boots 2X-Large
19-125-603 White Elastic wrist, ankle and back, AquaTrak™ boots 3X-Large
19-125-606 White Elastic wrist, ankle and back, AquaTrak™ boots 4X-Large
19-125-607 White Elastic wrist, ankle and back, AquaTrak™ boots 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
This item is not returnable. View return policy
Not recommended for use with chemicals.
Catalog No. 19-130-1502K Supplier AlphaProTech Supplier No. CV7403211
Add to Cart
Add to Cart
This item is not returnable. View return policy
Not recommended for use with chemicals.

Available in two styles and two colors, AlphaGuard™ offers a unique blend of strength, comfort, and protection in controlled environments.

  • Low particle count
  • Elastic back
  • Serged seams

Specifications

Product Type Cleanroom Coveralls
Size 8X-Large
Material SMS
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Seam Type Serged
Sterility Non-sterile
Back Style Elastic
Description Elastic wrist, ankle and back
For Use With (Application) For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency
Style AlphaGuard™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.