Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ GenPro™ Frocks
BIOPHARMA

Catalog No. 191301516A Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Length (Metric) Sleeve:
85.09 cm
87.63 cm
90.17 cm
92.71 cm
95.25 cm
101.6 cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (Metric) Sleeve
19-130-1516A Small 85.09 cm
19-130-1516B Medium 87.63 cm
19-130-1516C Large 90.17 cm
19-130-1516D X-Large 92.71 cm
19-130-1516F 3X-Large 101.6 cm
19-130-1516E 2X-Large 95.25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-130-1516A Supplier AlphaProTech Supplier No. FK121221
Add to Cart
Add to Cart
This item is not returnable. View return policy

Polypropylene frocks offers protection for non-hazardous environments. Ideal for facility visitors.

  • Polypropylene frock ideal for personnel or facility visitors
  • Offer comfort and breathability
  • Extra long design provides additional coverage
  • No pockets to help control contaminants from entering or leaving sensitive areas
  • Elastic wrist
  • Snap mandarin collar
  • Five-snap closure

Specifications

Size Small
Material Composite Fabric
Color White
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Snap Mandarin
Size (English) Chest 21.75 in.
Size (Metric) Chest 55.24 cm
Sleeve Style Raglan Sleeve
Length (Metric) Sleeve 85.09 cm
Product Type Controlled Environments Frock
Seam Type Serged
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.