Learn More
Moldex™ Alphas™ Reusable Ear Plugs
SDP
Catalog No. 19384362 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Corded:
Corded
Corded (detachable)
Uncorded
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corded
19-384-362 Corded (detachable)
19-384-363 Corded
19-384-361 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-384-362 Supplier Moldex™ Supplier No. 6434
Alphas™ reusable ear plugs unique ridged stem allows the user to get a solid grip on the ear plug for ease of insertion and removal, even when wearing gloves.
  • Economic and reusable
  • Corded and uncorded
  • Corded option with cloth cord
  • Pocket-Pak Plus provides hygienic storage of plugs between uses
  • Independently tested NRR 27
  • 100% PVC-free
Specifications
ANSI S3.1974 Yes
Cord Color Green
Cord Material Cloth
CSA Z94.2 Class AL
Size Universal
High Visibility No
Pairs per Package 50 Pairs/Pk.
Plug Material 100% PVC-free
Plug Color Gray
Noise Reduction Rating (NRR) 27 dB
Corded Corded (detachable)
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Includes Pocket-size case