Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves

Catalog No. 19010060 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-060 8
19-010-058 6
19-010-059 7
19-010-061 9
19-010-062 10
19-010-063 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-010-060 Supplier Ansell Supplier No. 988826
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.

  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort

 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN420:2003 + A1:2009; Category lll
Color White
Latex Free Yes
Material Laminated Film
Size 8
Style AlphaTec™ 02-100
Thickness (English) 2.5 mil
Thickness (Metric) 0.06 mm
EN388 Standard EN 388:0000
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCEFGHIMS
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Grip Design Smooth
Lining Unlined
Model 988826
Series AlphaTec™ 02-100
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.