Learn More
Ansell AlphaTec™ 1500 PLUS - Lab Coat Model 208
Catalog No. 17000256 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-256 Small
17-000-257 Medium
17-000-258 Large
17-000-260 -
17-000-262 4X-Large
17-000-263 5X-Large
17-000-259 X-Large
17-000-261 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-000-256 Supplier Ansell Supplier No. WH15B9220802
Only null left
Add to Cart
Add to Cart
Breathable lab coats are made of anti-static SMS material suitable for electro-static sensitive and manufacturing work environments.
  • Short style, white lab coats
  • SMS fabric construction
  • Anti-static treatment
  • Air and water vapor permeable
  • Silicone-free, bound seams and low-linting
  • Elastic cuffs, studded front-fastening and left-breast & right-thigh integral pockets
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Size Small
Gender Unisex
Material SMS
Color White
Closure Type Press Studs
Cuff Style Elastic
Resistance Static
Collar Type Serged
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [6])
For Use With (Application) Unpacking Bulk Goods, Loading and Unloading Processing Equipment, Production Line Support and Maintenance, Assembly and Inspection of Components
No. per Pack 70
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.