Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 1500 PLUS Stitched Model 111, Coveralls with Hood and Finger Loops

Catalog No. 17010445 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-445 X-Large
17-010-441 Small
17-010-442 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-010-445 Supplier Ansell Supplier No. WH15S9211105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Particle-filtering, anti-static SMS protective suit delivers light chemical resistance.

  • Particle-filtering SMS fabric: Filters 100% of particles over 3 microns(According to KAKEN particle penetration test)
  • Air and water vapor permeability: Ensures heat stress protection
  • Optimized body fit: Enhanced comfort
TRUSTED_SUSTAINABILITY

Specifications

Product Type Single-use Coveralls
Size X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Particle, Anti-static
Certifications/Compliance (Europe) CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN 1149-5)
Latex Free No
Model WH15-S-92-111
No. of Pockets None
For Use With (Application) Machinery and Equipment, Life Sciences, Metal Fabrication, Construction, Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.