Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 23-201 Long Sleeve PVC Glove

Catalog No. 19500760 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-760 10
19-500-757 7
19-500-758 8
19-500-759 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-500-760 Supplier Ansell Supplier No. 23201100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Long sleeve 24 in. PVC glove offers good dexterity and pliability, even at low temperatures. Dipped rough finish makes handling wet and slippery materials easier and safer.

  • Rough sandblast finish provide good wet-grip
  • Medium duty glove
  • 13 gauge knitted cotton liner
  • PVC sleeve at arm
  • Food approved
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA
Certifications/Compliance (Europe) EU Food Handling, EN 420:2003 + A1:2009; Category lll; REACH Compliant
Color Blue
Latex Free Yes
Material PVC
Product Type Glove
Size 10
Thickness (English) 63 mil
Thickness (Metric) 1.60 mm
EN388 Standard EN388:3121B, EN388:4121
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
For Use With (Application) Food Processing; Automotive; Oil and Gas
Glove Cuff Style Attached Sleeve
Grip Design Sandblast
Model 23201100
Length (English) 24 in.
Length (Metric) 62 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.