Learn More
Ansell™ AlphaTec™ 56-100 Heavy-Duty PVC Aprons
Catalog No. 19501041 Shop All Ansell Products
Change view
Click to view available options
Dimensions (L x W):
33 x 44 in.
Catalog No. Dimensions (L x W)
19-501-041 33 x 44 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-501-041 Supplier Ansell™ Supplier No. 950100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heavy duty PVS aprons with 18 mil die cut
  • Heavy duty PVC aprons - 18 mil die cut offer users a combination chemical splash protection and flexible comforts
  • 48 in. ties and sealed grommets provide a durable, consistent and adjustable fit on all sizes and styles
  • Protects from oil, grease, and chemicals
  • Durable
  • Offers resistance to abrasion and puncture 
  • Uses:
    • Food processing, Life sciences
  • TRUSTED_SUSTAINABILITY
    Specifications
    Material Vinyl
    Color Green
    Includes 12 Aprons/Carton
    Certifications/Compliance Cat III, EN 14605:2005, A1:2009 Type PB4 66 x 3326 X, EN ISO 13688:2013
    Dimensions (L x W) 33 x 44 in.
    Style AlphaTec 56-100 Vinyl PVC
    Adhere to
    Safety Regulations and Best Practices
    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.