Learn More
Ansell AlphaTec™ 58-435 Heavy Duty Nitrile Gloves
Catalog No. 17999820 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-820 11
17-999-816 7
17-999-817 8
17-999-818 9
17-999-819 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-999-820 Supplier Ansell Supplier No. 256252
  • $258.16
    $112.65 / Pack of 12
    Save $145.51
    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,414.63
    $617.65 / Case of 6 PK
    Save $796.98
    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heavy-duty nitrile gloves with ANSELL GRIP™ technology features a 16 mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters.
  • ANSELL GRIP™ Technology for optimal control when handling oily or wet objects
  • Premium grade nitrile shell offers excellent chemical protection against a wide range of chemicals
  • Lightweight, with cotton flocked lining offering superior comfort
  • Enhanced grip, dexterity and comfort helps reduce hand fatigue
  • Extra-long cuff extends protection to the forearm
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III, EN 1149
Chlorinated No
Color Gray
Cut Resistance Rating ANSI A5
Latex Free Yes
Material Nitrile
Size 11
EN388 Standard EN 388
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLOPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design ANSELL GRIP™ Technology
Liner Material Cotton Flock
Lining Lined
Model 256252
Series AlphaTec™ 58-435
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.