Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 62-400 Heavyweight Natural Rubber Supported Gloves

Catalog No. 19821916 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-916 8
19-821-917 9
19-821-918 10
19-821-919 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-821-916 Supplier Ansell Supplier No. 285652/DEL
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Flexible and durable 12 in. natural rubber gloves for wide range of applications requiring chemical protection.

  • Fully coated natural rubber latex glove
  • Soft-knit cotton liner helps keep hands and fingers comfortable in hot or cold environments
  • Stays flexible in cold temperatures
  • Heavy duty with rough crinkle finish
  • Ergonimically fit
  • Ideal for adjusting parts, systems and components.
  • Ideal for food processing applications that have food contact
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Color Blue
Latex Free No
Material Latex
Size 8
EN388 Standard EN 388:2131, EN 388:2131B
EN 421 No
EN ISO 374-1 Type C
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Pinked
Grip Design Crinkle Finish
Liner Material Cotton
Lining Lined
Model 62-400 8 104634
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.