Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 87-085 Natural Rubber Gloves

Catalog No. 17609984 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-609-984 10
17-609-981 7
17-609-982 8
17-609-983 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-609-984 Supplier Ansell Supplier No. 87085100BP
  • $655.17
    $416.65 / Case of 144

    Save $238.52

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flocked natural rubber gloves, offering elevated comfort and food handling compliance.

Key Features and Benefits

  • Beaded cuff: Easy-to-don, tear and chemical-resistant gloves
  • Food-safe: Ideal for processing food and protecting hands
  • Good abrasion resistance: Designed for use in low-hazard areas

Description

  • Increased protection and convenience: AlphaTec™ 87-085 rubber work gloves are flexible, lightweight and feature tear-resistant beaded cuffs for easy donning and doffing
  • Food handling compliance: As these unsupported protective gloves possess an AQL of 1.5, they are also suitable for food processing applications
  • Added durability: AlphaTec™ 87-085 abrasion-resistant gloves offer moderate durability when working in agriculture, food processing and chemical industries
  • Improved comfort: These pink safety gloves feature 100% cotton flock lining, which ensures flexibility and elevated comfort
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 374-1:2016, EN ISO 374-5:2016, Food Type: Rubber, Category III
Certifications/Compliance (Europe) CE 0493, EN 388:2016 +A1:2018, REACH Compliant
Chlorinated Yes
Applications Industrial, Laboratory
Color Pink
Finger Thickness (English) 18 mil
Finger Thickness (Metric) 0.45 mm
Latex Free No
Material Rubber Latex
Palm Thickness (English) 18 mil
Palm Thickness (Metric) 0.45 mm
Product Type Chemical Protection Gloves
Size 10
Thickness (English) 18 mil
Thickness (Metric) 0.45 mm
EN388 Standard EN 388:2016 +A1:2018 - 2010X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Beaded
Grip Design Raised Diamond
Lining Unlined
Length (English) 11.8 in.
Length (Metric) 300 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.