Learn More
Ansell AlphaTec™ BSL4 Orange Protective Suit with Attached PVC Safety Boots
Catalog No. 17100999 Shop All Ansell Products
Change view
Click to view available options
Size:
2X-Large (Suit), 12 (Boots)
2X-Large (Suit), 13 (Boots)
2X-Large (Suit), 14 (Boots)
Large (Suit), 10 (Boots)
Large (Suit), 11 (Boots)
Large (Suit), 12 (Boots)
Large (Suit), 9 (Boots)
Medium (Suit), 10 (Boots)
Medium (Suit), 7 (Boots)
Medium (Suit), 8 (Boots)
Medium (Suit), 9 (Boots)
Small (Suit), 6 (Boots)
Small (Suit), 7 (Boots)
Small (Suit), 8 (Boots)
X-Large (Suit), 11 (Boots)
X-Large (Suit), 12 (Boots)
X-Large (Suit), 13 (Boots)
X-Large (Suit), 14 (Boots)
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-100-999 Large (Suit), 12 (Boots)
17-100-989 Small (Suit), 6 (Boots)
17-100-990 Small (Suit), 7 (Boots)
17-100-991 Small (Suit), 8 (Boots)
17-100-992 Medium (Suit), 7 (Boots)
17-100-993 Medium (Suit), 8 (Boots)
17-100-994 Medium (Suit), 9 (Boots)
17-100-995 Medium (Suit), 10 (Boots)
17-100-996 Large (Suit), 9 (Boots)
17-100-997 Large (Suit), 10 (Boots)
17-100-998 Large (Suit), 11 (Boots)
17-101-000 X-Large (Suit), 11 (Boots)
17-101-001 X-Large (Suit), 12 (Boots)
17-101-002 X-Large (Suit), 13 (Boots)
17-101-003 X-Large (Suit), 14 (Boots)
17-101-004 2X-Large (Suit), 12 (Boots)
17-101-005 2X-Large (Suit), 13 (Boots)
17-101-006 2X-Large (Suit), 14 (Boots)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 17-100-999 Supplier Ansell Supplier No. ALOBNC696000012004
  • $9,865.00
    $7,691.65 / Each of 1
    Save $2,173.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Soft, flexible and durable airfed suit designed for combined chemical, biological and respiratory protection.
  • Suitable for Level 4 Biological Safety Laboratories
  • Provides protection against chemicals in liquid, aerosol and solid form
  • Protects against infectious agents, i.e. bacteria, virus and fungi, and against radioactive particles
  • Designed for use with an external air source, which allows longer working hours and provides improved wearer comfort
  • Made from a strong yet soft and durable material
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
For Use With (Application) Biosafety Labs Levels 1-4, Centers for Disease Control and Prevention, Life Sciences, Chemical, Mining, Fire and Rescue, Oil and Gas
Material Polyester
Size Large (Suit), 12 (Boots)
Closure Type Heavy-duty vapor-protective zipper with D-rings
Disposable Reusable
Product Type Chemical Protective Suits
Seam Type Stitched and welded