Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ Medium-Duty Chemical Protection Gloves

Catalog No. 19500892 Shop All Ansell Products
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-892 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-500-892 Supplier Ansell Supplier No. 55305100/DEL

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AnsellAlphaTec™ Medium-Duty Chemical Protection Glove delivers chemical resistance against most acids, alcohols, oils, lubricants and hydrocarbons

  • Strong physical protection against abrasions and good protection against cuts
  • Medium thickness increases dexterity, which improves the ability to manipulate objects
  • Resistant to Type I allergies
  • Unlined for effective cleaning
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 0
Color Black
Latex Free Yes
Material Neoprene
Size 10
Thickness (Metric) 0.51 mm
EN388 Standard EN 388:20032100; EN 388:20162100A
EN 421 No
EN ISO 374-1 Type A
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Series AlphaTec™ 55-305
Length (English) 32 in.
Length (Metric) 81.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.