Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 2000 Comfort Bound Coveralls with Hood, Model 129

Catalog No. 17999891 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-891 5X-Large
17-999-884 Small
17-999-886 Large
17-999-885 Medium
17-999-887 X-Large
17-999-888 2X-Large
17-999-889 3X-Large
17-999-890 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-999-891 Supplier Ansell Supplier No. WH20B9212909
  • $723.90
    $372.65 / Case of 25

    Save $351.25

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coveralls protective barrier shields hood, arms, legs, and front torso against particulates, liquids, and light chemical splash. Three-piece hood ensures adequate coverage and finger loops improve comfort and usability. Previously known as MICROCHEM™ by AlphaTec 2000 Comfort.

  • Arms, legs, and front torso in 2000 fabric
  • Protects against liquid, light chemical resistance,and particulate biological agents
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free
  • Anti-static, tested according to EN1149-5 and AATCC76
  • Elastic ankles and wrists
  • 3-piece hood, 2-way front zipper with storm flap
  • Bound seams, finger loops
  • Ideal for pharmaceutical industries, cleanrooms, paint spraying, veterinary services, pest control
  • Available in Small through 5X-Large
  • Certifications EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 5X-Large
Gender Unisex
Material Microporous Laminate, SMS
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid, Chemicals
Back Style Breathable SMS Back Panel
Certifications/Compliance AATTC 76 Compliant
Certifications/Compliance (Europe) EN 1149-5, CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN 1149-5)
Latex Free Yes
Model WH20-B-92-129
For Use With (Application) Automotive, Life Sciences, Metal Fabrication, Veterinary, Maintenance, Cleaning, Production, Food Processing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.