Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Hood and Socks

Catalog No. 17999917 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-917 Medium
17-999-916 Small
17-999-918 Large
17-999-919 X-Large
17-999-920 2X-Large
17-999-921 3X-Large
17-999-923 5X-Large
17-999-922 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17-999-917 Supplier Ansell™ Supplier No. YY23B9214703
Add to Cart
Add to Cart
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.

Provide an excellent barrier to harmful chemicals and blood borne pathogens. Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Hood and Socks are lightweight and relatively strong and durable. These coveralls are designed for applications such as environmental clean-up, sewage purification installations, and composites manufacturing.

  • Protection:  A barrier to numerous inorganic liquid chemicals including acids and bases; achieves the highest classifications for protection from biological agents in accordance with EN 14126:2003 & ASTM F1671 for penetration of blood, bodily fluids, and blood-borne pathogens
  • Comfort:  Lightweight yet strong and durable
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, bound seams, finger loops, 3 piece respirator fit hood, attached socks
  • Applications:  Environmental cleanup, sewage purification installations, composites manufacturing
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671

TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Waist Style Elastic
Ankle Style Elastic, Attached Socks
Cuff Style Elastic, Thumb Loop
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Bound
Protection Class Particulate and light liquid splash protection
Latex Free No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.