Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood and Socks

Catalog No. 17999948 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-948 Small
17-999-949 Medium
17-999-950 Large
17-999-951 X-Large
17-999-952 2X-Large
17-999-953 3X-Large
17-999-954 4X-Large
17-999-955 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17-999-948 Supplier Ansell™ Supplier No. OR50T9212202
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions.

Provide strong, durable protection suitable for workers in extremely hazardous areas, including HAZMAT reponse teams. Ansell™ Alphatec™ MICROCHEM™ 68-5000 Orange Coveralls with Hood and Socks reach new levels in chemical protection and have been engineered to protect.

  • Protection:  Barrier to numerous organic and inorganic chemicals and biological hazards; >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Comfort:  Multi-layer material which is lightweight, yet strong and durable
  • Highly visible:  Bright orange color for improved worker safety
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece hood, integrated socks with boot overflap
  • Applications:  Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, mining, agriculture, HAZMAT emergency reponse (Level B), first response and fire service
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

TRUSTED_SUSTAINABILITY

Specifications

Size Small
Material Multi-layer Barrier Laminate Fabric
Color Orange
Waist Style Elastic
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic, Knit
Neck Style Hooded
Hood Type 2-panel
Closure Type Double Zipper
Seam Type Welded, Taped
Protection Class Liquid and spray tight chemical and biological protection
Description Elasticated outer cuff with knitted inner cuff
Latex Free No
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.