Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Attached Gloves

Catalog No. 18000020 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-000-020 Small
18-000-021 Medium
18-000-022 Large
18-000-023 X-Large
18-000-025 3X-Large
18-000-024 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 18-000-020 Supplier Ansell™ Supplier No. OR60T9280102G02
This item is not returnable. View return policy

Provide protection for emergency responders or chemical workers against dangerous and toxic chemicals in liquid or gaseous form. Ansell™ Alphatec™ MICROCHEM™ 68-6000 Gastight Coveralls with Attached Gloves are Level A suits where a self-contained breathing apparatus is worn on the inside.

  • Protection:  Level A Gas tight suit tested against numerous chemicals including the ASTM F1001 test battery
  • Fabric technology:  Co-extruded multi-layer performance barrier laminate with scrim reinforcement
  • Visor technology:  Semi-rigid 3 layer PET visor providing excellent optical clarity
  • Design:  DYNAT/YKK gas-tight zipper, permanently attached Ansell Barrier gloves with sleeve over-cuffs, ultrasonically welded and taped seams
  • Applications:  Chemicals, pharmaceuticals, emergency service, sewage treatment, nuclear, petrochemicals, first response, industrial and tank cleaning, shipping
  • Sizes:  Available in Small through 3X-Large
  • Certifications:  EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN1149-5 / AATCC 76, EN 14126 / ASTM F1671, EN943-1 Type 1a, EN943-2 Type 1a-ET

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Material Barrier Laminate Fabric
Size Small
Ankle Style Attached socks, Boot overflaps
Closure Type Gas-tight zipper
Cuff Style Elastic, Knit
Description Permanently attached Ansell Barrier glove with sleeve over-cuffs
Protection Class Gas tight chemical and biological protection
Seam Type Welded, Taped

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.