Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-CFR Flame Retardant Coveralls with Hood

Catalog No. 17999956 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-956 Small
17-999-957 Medium
17-999-958 Large
17-999-959 X-Large
17-999-960 2X-Large
17-999-962 4X-Large
17-999-961 3X-Large
17-999-963 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.
Catalog No. 17-999-956 Supplier Ansell™ Supplier No. RD96T9211102
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
This product contains natural rubber latex which may cause allergic reactions. Must be worn over thermal protective garments, such as NOMEX™ or PYROVATEX™, and should never be worn next to the skin.

Offer protection from particulates and chemical sprays or splashes without compromising worker protection in the event of a flashfire. Ansell™ Alphatec™ MICROCHEM™ 68-CFR Flame Retardant Coveralls are manufactured from flame retardant and antistatic fabric. They are ideal for use in industries like oil and petrochemicals, petrolium distribution and processing, and utilities.

  • Protection:  Flame retardant treated fabric with PVC barrier film offering wearers protection from liquid chemicals
  • Versatile:  Good for most applications where there is the need for protection from chemical spray without compromising wearer protection in the event of a flash fire
  • Optimized body fit:  Improves wearer comfort and safety
  • Highly visible:  Bright red color to improve worker safety
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, serged and taped seams, 2 piece hood
  • Applications:  Oil and petrochemicals, petroleum distribution and processing, utilities
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 14116 INDEX 1/0/0
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, EN 14126 / ASTM F1671, EN 14605 Type 3, EN 14605 Type 4, EN ISO 14116 INDEX 1/0/0

TRUSTED_SUSTAINABILITY

Specifications

Size Small
Material CFR
Color Red
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Closure Type Double Zipper
Seam Type Serged, Taped
Protection Class Liquid and spray tight chemical and biological protection
Latex Free No
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.