Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ AlphaTec™ Overboots

Catalog No. 19010379 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-379 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-010-379 Supplier Ansell™ Supplier No. GR40W9240600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ansell™ AlphaTec™ Overboots feature multi-layer chemical barrier technology

  • Over 200 chemicals permeation tested, including chemical warfare agents
  • Silicone free
  • Applications:
    • Contaminated environments
    • Chemical handling
    • Filling, blending and charging of raw materials
    • Transferring liquids and solids
    • Opening and draining pumps valves or lines
    • Loading and unloading of process equipment
    • Raw material sample collection
    • Concentration mixing and palletisation
    • Hazmat response
    • Assembly and disassembly of large pipes
TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Material Laminate
Color Green
Closure Type Elastic with Ties
Seam Type Welded seams
Certifications/Compliance CE CATEGORY I
Weight (Metric) 103 gsm

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.