Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ AlphaTec™ Protective Bib Apron

Catalog No. 19010370 Shop All Ansell Products
Change view
Click to view available options
Weight (Metric):
103 gsm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
19-010-370 103 gsm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-010-370 Supplier Ansell™ Supplier No. GR40W9221200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ansell™ AlphaTec™ Protective Bib Apron features multi-layer chemical barrier technology.

  • Over 200 chemicals permeation tested, including chemical warfare agents
  • Silicone free
  • Applications:
    • Contaminated environments
    • Chemical handling
    • Filling, blending and charging of raw materials
    • Transferring liquids and solids
    • Opening and draining pumps valves or lines
    • Loading and unloading of process equipment
    • Raw material sample collection
    • Concentration mixing and palletisation
    • Hazmat response
    • Assembly and disassembly of large pipes
TRUSTED_SUSTAINABILITY

Specifications

Type Protective Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Green
Closure Type Ties
Sterility Non-sterile
Construction Welded Seams
Certifications/Compliance CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126)
Dimensions (L x W) 28 x 40 in.
Weight (Metric) 103 gsm
Style Bib apron
Seam Type Welded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.